Assay Details
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177506.2 | NP_803472.1 | 4 - 5 | 812 | 84 | |
GenBank mRNA | BC120315.1 | - | 4 - 5 | 731 | 84 | |
M80626.1 | - | 4 - 5 | 812 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
