Assay Details
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177506.2 | NP_803472.1 | 5 - 6 | 907 | 76 | |
GenBank mRNA | BC120315.1 | - | 5 - 6 | 826 | 76 | |
M80626.1 | - | 5 - 6 | 907 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
