Assay Details
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177512.2 | NP_803478.1 | 1 - 2 | 610 | 102 | |
GenBank mRNA | AF515786.2 | - | 1 - 2 | 208 | 102 | |
BC120415.1 | - | 1 - 2 | 579 | 102 | ||
J05217.1 | - | 1 - 2 | 450 | 102 | ||
M13214.1 | - | 1 - 2 | 225 | 102 | ||
X14558.1 | - | 610 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
