Assay Details
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180993.2 | NP_851336.1 | 2 - 3 | 806 | 95 | |
GenBank mRNA | BC102820.1 | - | 3 - 4 | 829 | 95 | |
J02659.1 | - | 2 - 3 | 807 | 95 | ||
M27606.1 | - | 2 - 3 | 820 | 95 | ||
Y00405.1 | - | 840 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
