Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180997.2 | NP_851340.2 | 1 - 2 | 195 | 117 | |
GenBank mRNA | AF348423.1 | - | 1 - 2 | 148 | 117 | |
EU276068.1 | - | 1 - 2 | 155 | 117 | ||
FE004067.1 | - | 1 - 2 | 195 | 117 | ||
FE016063.1 | - | 3 - 4 | 605 | 117 | ||
M12791.1 | - | 1 - 2 | 165 | 117 | ||
M13204.1 | - | 1 - 2 | 178 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
