Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180997.2 | NP_851340.2 | 2 - 3 | 254 | 106 | |
GenBank mRNA | AF348423.1 | - | 2 - 3 | 106 | ||
EF067918.1 | - | 2 - 3 | 218 | 106 | ||
EU276068.1 | - | 2 - 3 | 214 | 106 | ||
FE004067.1 | - | 2 - 3 | 254 | 106 | ||
FE016063.1 | - | 2 - 3 | 546 | 106 | ||
M12791.1 | - | 2 - 3 | 224 | 106 | ||
M13204.1 | - | 2 - 3 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
