Assay Details
Gene Symbol: FAHD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205790.1 | NP_991359.1 | 1 - 2 | 166 | 82 | |
GenBank mRNA | AY192436.1 | - | 1 - 2 | 166 | 82 | |
BC112705.1 | - | 1 - 2 | 156 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112705.1, NM_205790.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
