Assay Details
Gene Symbol: FAHD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205790.1 | NP_991359.1 | 7 - 8 | 1051 | 79 | |
GenBank mRNA | AY192436.1 | - | 7 - 8 | 1051 | 79 | |
BC112705.1 | - | 7 - 8 | 1041 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
