Assay Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206969.1 | NP_996852.1 | 1 - 2 | 76 | 69 | |
GenBank mRNA | AY554049.1 | - | 1 - 2 | 76 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_206969.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
