Assay Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206969.1 | NP_996852.1 | 6 - 7 | 627 | 101 | |
GenBank mRNA | AY554049.1 | - | 6 - 7 | 627 | 101 | |
BC149481.1 | - | 4 - 5 | 799 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
