Assay Details
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012670.2 | NP_001012688.1 | 2 - 3 | 310 | 66 | |
GenBank mRNA | AB072368.1 | - | 2 - 3 | 277 | 66 | |
BC102618.1 | - | 2 - 3 | 225 | 66 | ||
BC151818.1 | - | 2 - 3 | 226 | 66 | ||
EV615064.1 | - | 2 - 3 | 314 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
