Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013592.1 | NP_001013610.1 | 5 - 6 | 524 | 64 | |
GenBank mRNA | AB098974.1 | - | 4 - 5 | 63 | ||
BC104579.1 | - | 4 - 5 | 511 | 64 | ||
BT021005.1 | - | 5 - 6 | 524 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
