Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013592.1 | NP_001013610.1 | 8 - 9 | 1050 | 97 | |
GenBank mRNA | BC104579.1 | - | 7 - 8 | 1037 | 97 | |
BT021005.1 | - | 8 - 9 | 1050 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
