Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014849.1 | NP_001014849.1 | 5 - 6 | 640 | 68 | |
NM_001113241.1 | NP_001106712.1 | 5 - 6 | 640 | 68 | ||
GenBank mRNA | BC109578.1 | - | 5 - 6 | 604 | 68 | |
BT021836.1 | - | 5 - 6 | 658 | 68 | ||
DT837410.1 | - | 5 - 6 | 641 | 68 | ||
DT847731.1 | - | 4 - 5 | 374 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
