Assay Details
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014925.1 | NP_001014925.1 | 5 - 6 | 896 | 123 | |
GenBank mRNA | BC119894.1 | - | 3 - 4 | 844 | 123 | |
BT021863.1 | - | 5 - 6 | 896 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
