Assay Details
Gene Symbol: MFSD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014925.1 | NP_001014925.1 | 7 - 8 | 1118 | 105 | |
GenBank mRNA | BC119894.1 | - | 5 - 6 | 1066 | 105 | |
BT021863.1 | - | 7 - 8 | 1118 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
