Assay Details
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015543.2 | NP_001015543.1 | 1 - 2 | 27 | 76 | |
GenBank mRNA | AY911336.1 | - | 1 - 2 | 28 | 76 | |
BC102865.1 | - | 1 - 2 | 31 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102865.1, NM_001015543.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
