Assay Details
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015555.1 | NP_001015555.1 | 1 - 2 | 171 | 118 | |
GenBank mRNA | BC102888.1 | - | 1 - 2 | 117 | 118 | |
BT021065.1 | - | 1 - 2 | 171 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102888.1, NM_001015555.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
