Assay Details
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015577.1 | NP_001015577.1 | 1 - 2 | 174 | 97 | |
GenBank mRNA | BT020881.1 | - | 1 - 2 | 174 | 97 | |
BT021707.1 | - | 1 - 2 | 165 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
