Assay Details
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015592.1 | NP_001015592.1 | 1 - 2 | 281 | 59 | |
GenBank mRNA | BC102240.1 | - | 1 - 2 | 293 | 59 | |
BT021078.1 | - | 1 - 2 | 281 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
