Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015601.1 | NP_001015601.1 | 5 - 6 | 695 | 56 | |
GenBank mRNA | BT020843.1 | - | 5 - 6 | 695 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
