Assay Details
Gene Symbol: CLIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015608.1 | NP_001015608.1 | 3 - 4 | 448 | 98 | |
GenBank mRNA | BC102103.1 | - | 3 - 4 | 299 | 98 | |
BT021003.1 | - | 3 - 4 | 448 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
