Assay Details
Gene Symbol: CLIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015608.1 | NP_001015608.1 | 4 - 5 | 558 | 153 | |
GenBank mRNA | AY999167.1 | - | 1 - 2 | 82 | 153 | |
BC102103.1 | - | 4 - 5 | 409 | 153 | ||
BT021003.1 | - | 4 - 5 | 558 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
