Assay Details
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015636.2 | NP_001015636.2 | 12 - 13 | 1403 | 72 |
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT020935.1 | - | 8 - 9 | 1204 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
