Assay Details
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015636.2 | NP_001015636.2 | 13 - 14 | 1497 | 76 |
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT020935.1 | - | 9 - 10 | 1298 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
