Assay Details
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015636.2 | NP_001015636.2 | 14 - 15 | 1529 | 96 |
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT020935.1 | - | 10 - 11 | 1330 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
