Assay Details
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015636.2 | NP_001015636.2 | 18 - 19 | 1832 | 84 |
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT020935.1 | - | 14 - 15 | 1633 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
