Assay Details
Gene Symbol: VPS72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015658.1 | NP_001015658.1 | 4 - 5 | 618 | 86 | |
GenBank mRNA | BC109639.1 | - | 4 - 5 | 583 | 86 | |
BT020964.1 | - | 4 - 5 | 618 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
