Assay Details
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017936.1 | NP_001017936.1 | 3 - 4 | 658 | 68 | |
GenBank mRNA | BC120091.1 | - | 3 - 4 | 658 | 68 | |
BT021481.1 | - | 3 - 4 | 658 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
