Assay Details
Gene Symbol: RPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024568.2 | NP_001019739.1 | 2 - 3 | 205 | 78 | |
GenBank mRNA | AY911366.1 | - | 2 - 3 | 206 | 78 | |
BC102249.1 | - | 2 - 3 | 172 | 78 | ||
MH936678.1 | - | 2 - 3 | 154 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
