Assay Details
Gene Symbol: GPX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025335.2 | NP_001020506.2 | 1 - 2 | 116 | 128 | |
GenBank mRNA | DQ058076.1 | - | 1 - 2 | 116 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
