Assay Details
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025339.2 | NP_001020510.1 | 1 - 2 | 39 | 69 | |
GenBank mRNA | AY911383.1 | - | 1 - 2 | 40 | 69 | |
BC103133.1 | - | 1 - 2 | 38 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103133.1, NM_001025339.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
