Assay Details
Gene Symbol: C17H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031753.1 | NP_001026923.1 | 5 - 6 | 460 | 110 | |
GenBank mRNA | BT021897.1 | - | 5 - 6 | 460 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
