Assay Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034038.1 | NP_001029210.1 | 1 - 2 | 93 | 107 | |
GenBank mRNA | AY911372.1 | - | 1 - 2 | 93 | 107 | |
BC102789.1 | - | 1 | 89 | 107 | ||
BC151408.1 | - | 1 | 93 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
