Assay Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034229.1 | NP_001029401.1 | 1 - 2 | 77 | 90 | |
GenBank mRNA | BC123409.1 | - | 1 - 2 | 126 | 90 | |
BT021734.1 | - | 1 - 2 | 77 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123409.1, NM_001034229.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
