Assay Details
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034233.2 | NP_001029405.1 | 3 - 4 | 410 | 95 | |
GenBank mRNA | AY911343.1 | - | 3 - 4 | 413 | 95 | |
BC103328.1 | - | 3 - 4 | 411 | 95 | ||
BC151811.1 | - | 3 - 4 | 428 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
