Assay Details
Gene Symbol: ECI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034242.2 | NP_001029414.2 | 4 - 5 | 551 | 70 | |
GenBank mRNA | BC102906.1 | - | 4 - 5 | 550 | 70 | |
EH139300.1 | - | 4 - 5 | 551 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
