Assay Details
Gene Symbol: ICOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034275.1 | NP_001029447.1 | 1 - 2 | 66 | 142 | |
GenBank mRNA | BC149958.1 | - | 1 - 2 | 81 | 142 | |
BT021638.1 | - | 1 - 2 | 66 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
