Assay Details
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034276.1 | NP_001029448.1 | 1 - 2 | 236 | 118 | |
GenBank mRNA | BC103180.1 | - | 1 - 2 | 236 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
