Assay Details
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034285.2 | NP_001029457.1 | 1 - 2 | 93 | 103 | |
GenBank mRNA | BC102636.1 | - | 1 - 2 | 94 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102636.1, NM_001034285.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
