Assay Details
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034389.1 | NP_001029561.1 | 1 - 2 | 56 | 73 | |
GenBank mRNA | BC103244.1 | - | 1 - 2 | 56 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103244.1, NM_001034389.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
