Assay Details
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034405.2 | NP_001029577.1 | 2 - 3 | 439 | 150 | |
GenBank mRNA | BC103441.1 | - | 2 - 3 | 440 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
