Assay Details
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034405.2 | NP_001029577.1 | 4 - 5 | 1316 | 147 | |
GenBank mRNA | BC103441.1 | - | 4 - 5 | 1317 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
