Assay Details
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034423.2 | NP_001029595.1 | 1 - 2 | 185 | 71 | |
GenBank mRNA | BC103437.1 | - | 1 - 2 | 186 | 71 | |
BT029898.1 | - | 1 - 2 | 152 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
