Assay Details
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034423.2 | NP_001029595.1 | 4 - 5 | 463 | 79 | |
GenBank mRNA | BC103437.1 | - | 4 - 5 | 464 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
