Assay Details
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034423.2 | NP_001029595.1 | 5 - 6 | 545 | 82 | |
GenBank mRNA | BC103437.1 | - | 5 - 6 | 546 | 82 | |
BT029898.1 | - | 5 - 6 | 512 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
