Assay Details
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034478.1 | NP_001029650.1 | 4 - 5 | 368 | 91 | |
GenBank mRNA | BC103304.1 | - | 4 - 5 | 368 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
