Assay Details
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034507.2 | NP_001029679.1 | 3 - 4 | 393 | 129 | |
GenBank mRNA | BC102506.1 | - | 3 - 4 | 394 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
