Assay Details
Gene Symbol: PAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034636.2 | NP_001029808.1 | 3 - 4 | 499 | 132 | |
GenBank mRNA | BC102885.1 | - | 3 - 4 | 501 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
