Assay Details
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034731.1 | NP_001029903.1 | 1 - 2 | 81 | 99 | |
GenBank mRNA | BC112634.1 | - | 1 - 2 | 70 | 99 | |
BT021483.1 | - | 1 - 2 | 81 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112634.1, NM_001034731.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
